SY3H3249F1BB Synthonix Corporation

1-Boc-pyrrolidine-3-ethanol

  • MDL number MFCD09833160
  • Cas Number : 160132-54-7
  • Molecular Weight : 215.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/1657.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/1657.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart