SY3H3249F1C5 Synthonix Corporation

1-BOC-azepane-4-carboxylic acid

  • MDL number MFCD10687156
  • Cas Number : 868284-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/1766.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/1766.html
hstatements H315 , H317 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart