SY3H3249F1C7 Synthonix Corporation

3-Pentyn-2-ol

  • MDL number MFCD00046652
  • Cas Number : 27301-54-8
  • Molecular Weight : 84.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/1790.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/1790.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart