SY3H3249F1CC Synthonix Corporation

tert-butyl N-(2-oxopiperidin-4-yl)carbamate

  • MDL number MFCD17170058
  • Cas Number : 1263281-78-2
  • Molecular Weight : 214.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/18307.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/18307.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart