SY3H3249F1D3 Synthonix Corporation

tert-butyl 7-(hydroxymethyl)-5-oxa-2-azaspiro[3.4]octane-2-carboxylate

  • MDL number MFCD20230641
  • Cas Number : 1330764-06-1
  • Molecular Weight : 243.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form oil
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/18684.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/18684.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart