SY3H3249F1F0 Synthonix Corporation

(1r,4r,5r)-rel-tertbutyl 5hydroxy2azabicyclo[2.2.1]heptane2carboxylate

  • Cas Number : 198835-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/22147.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/22147.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart