SY3H3249F293 Synthonix Corporation

(5-Bromo-2-methoxyphenyl)methanamine

  • MDL number No MDL found
  • Cas Number : 166530-78-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/38149.html

Safety Information

MSDS https://synthonix.com/38149.html
Availability details for
Quantity
  • Availability
  • Add to Cart