SY3H3249F34A Synthonix Corporation

2-Bromo-3,4-difluoro-1-nitrobenzene

  • MDL number MFCD01820590
  • Cas Number : 350699-92-2
  • Molecular Weight : 237.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Colorless to Pale-yellow to Yellow-brown Solid or Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/38950.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/38950.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart