SY3H3249F352 Synthonix Corporation

(R)-2-(((Benzyloxy)carbonyl)amino)-2-cyclohexylacetic acid

  • MDL number MFCD00191082
  • Cas Number : 69901-85-5
  • Molecular Weight : 291.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form powder Colour: white - off White
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/38986.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/38986.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart