SY3H3249F37C Synthonix Corporation

3-Bromo-5-cyclohexyl-4,5-dihydroisoxazole

  • MDL number MFCD11707002
  • Cas Number : 1120214-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/5020.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/5020.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart