SY3H3249F37D Synthonix Corporation

3-Bromo-5-phenyl-4,5-dihydroisoxazole

  • MDL number MFCD11707003
  • Cas Number : 86256-88-4
  • Molecular Weight : 226.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 93
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/5021.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/5021.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart