SY3H3249F37E Synthonix Corporation

3-Bromo-4,5-dihydroisoxazole-5-carboxylic acid amide

  • MDL number MFCD11707008
  • Cas Number : 1030613-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/5026.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/5026.html
hstatements H301 , H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart