SY3H3249F37F Synthonix Corporation

6-bromo-3,4-dihydro-2H-thiochromen-4-amine hydrochloride

  • MDL number MFCD09835500
  • Cas Number : 1170470-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/5119.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/5119.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart