SY3H3249F380 Synthonix Corporation

6-Bromo-3,4-dihydro-2H-S,S-dioxo-thiochromen-4-amine hydrochloride

  • MDL number MFCD11616744
  • Cas Number : 1172986-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://synthonix.com/5120.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/5120.html
hstatements H301 , H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart