SY3H3249F382 Synthonix Corporation

6-(tert-Butoxycarbonyl)-5,6,7,8-tetrahydropyrido[3,4-b]pyrazine-7-carboxylic acid

  • MDL number MFCD04973511
  • Cas Number : 264623-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/5379.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/5379.html
hstatements H315 , H317 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart