SY3H3249F385 Synthonix Corporation

2-Bromo-1,1-diethoxypropane

  • MDL number MFCD00152299
  • Cas Number : 3400-55-3
  • Molecular Weight : 211.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 94
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/5672.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/5672.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart