SY3H3249F38B Synthonix Corporation

2-Bromo-3-phenylpropanoyl chloride

  • MDL number MFCD19443915
  • Cas Number : 42762-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link https://synthonix.com/6285.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6285.html
hstatements H302 , H314 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart