SY3H3249F390 Synthonix Corporation

2-Benzhydryl-2-azaspiro[3.3]heptan-5-ol

  • MDL number MFCD20444452
  • Cas Number : 1263296-81-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/6694.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/6694.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart