SY3H3249F391 Synthonix Corporation

2-(tert-Butoxycarbonyl)-5-thia-2-azaspiro[3.4]octane-8-carboxylic acid 5,5-dioxide

  • MDL number MFCD22199158
  • Cas Number : 1340481-81-3
  • Molecular Weight : 305.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/6698.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/6698.html
pictograms GHS07
hstatements H301, H315, H317, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart