SY3H3249F393 Synthonix Corporation

tert-Butyl 4-(aminomethyl)-3,3-difluoropiperidine-1-carboxylate

  • MDL number MFCD18909808
  • Cas Number : 1303973-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/6840.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6840.html
hstatements H302 , H315 , H318 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart