SY3H3249F394 Synthonix Corporation

tert-Butyl 3,3-difluoro-4-(hydroxymethyl)piperidine-1-carboxylate

  • MDL number MFCD18909777
  • Cas Number : 1303974-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/6841.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6841.html
hstatements H302 , H315 , H318 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart