SY3H3249F395 Synthonix Corporation

tert-Butyl 3-bromopyrrolidine-1-carboxylate

  • MDL number MFCD08752478
  • Cas Number : 939793-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/6964.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6964.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart