SY3H3249F39D Synthonix Corporation

3-Bromodihydro-2H-pyran-4(3H)-one

  • MDL number MFCD13193743
  • Cas Number : 98021-79-5
  • Molecular Weight : 179.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7711.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/7711.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart