SY3H3249F39E Synthonix Corporation

Benzyl [2-(3,5-Bis(trifluoromethyl)phenyl)-1-methyl-2-oxo-ethyl]carbamate

  • MDL number MFCD19707642
  • Cas Number : 1363382-26-6
  • Molecular Weight : 419.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7761.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/7761.html
pictograms GHS05, GHS07
hstatements H302, H315, H318, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart