SY3H3249F3A1 Synthonix Corporation

cis-tert-Butyl octahydro-1H-pyrrolo[2,3-c]pyridine-1-carboxylate (rel)

  • MDL number MFCD13189556
  • Cas Number : 949559-11-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7880.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7880.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart