SY3H3249F3A5 Synthonix Corporation

1-Boc-3-fluoropiperidine-3-carboxylic acid

  • MDL number MFCD11847195
  • Cas Number : 934342-39-9
  • Molecular Weight : 247.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/8016.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/8016.html
pictograms GHS05, GHS07
hstatements H302, H315, H317, H318, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart