SY3H3249F3A7 Synthonix Corporation

1-Boc-3-(aminomethyl)-3-hydroxypyrrolidine

  • MDL number MFCD18917057
  • Cas Number : 114214-73-2
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/8056.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/8056.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart