SY3H3249F3AA Synthonix Corporation

4-(Boc-amino)-3,3-difluoropiperidine

  • MDL number MFCD18791569
  • Cas Number : 1263180-22-8
  • Molecular Weight : 236.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/8069.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/8069.html
pictograms GHS05, GHS07
hstatements H302, H315, H318, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart