SY3H3249F3AB Synthonix Corporation

1-Boc-3-aminomethyl-3-hydroxypiperidine

  • MDL number MFCD18917047
  • Cas Number : 1308384-31-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/8074.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/8074.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart