SY3H3249F3AC Synthonix Corporation

5-Amino-1-methylpiperidin-2-one hydrochloride

  • MDL number MFCD09864348
  • Cas Number : 1228838-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/0588.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/0588.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart