SY3H3249F3AE Synthonix Corporation

2-Bromomethyl-4,4-dimethyloxetane

  • MDL number MFCD22628725
  • Cas Number : 1408074-88-3
  • Molecular Weight : 179.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/8433.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/8433.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart