SY3H3249F3AF Synthonix Corporation

2-(Boc-amino)-2-(oxetan-3-yl)acetic acid

  • MDL number MFCD21848380
  • Cas Number : 1408074-43-0
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/8436.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/8436.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart