SY3H3249F3B1 Synthonix Corporation

2-Bromo-3-phenylpropionic acid

  • MDL number MFCD00461796
  • Cas Number : 16503-53-0
  • Molecular Weight : 229.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/8639.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/8639.html
pictograms GHS07
hstatements H315, H317, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart