SY3H3249F3C8 Synthonix Corporation

3-Cyano-4-oxo-piperidine-1-carboxylic acid tert-butyl ester

  • MDL number No MDL found
  • Cas Number : 914988-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/11959.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/11959.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart