SY3H3249F3D4 Synthonix Corporation

1-(4-chloropyridin-3-yl)ethan-1-one

  • MDL number No MDL found
  • Cas Number : 116922-73-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/16933.html

Safety Information

MSDS https://synthonix.com/16933.html
Availability details for
Quantity
  • Availability
  • Add to Cart