SY3H3249F3FB Synthonix Corporation

3,4-Dihydro-1H-isochromen-4-amine hydrochloride

  • MDL number MFCD08448171
  • Cas Number : 147663-00-1
  • Molecular Weight : 185.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/4046.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/4046.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart