SY3H3249F401 Synthonix Corporation

N-Hydroxysulfosuccinimide sodium salt

  • MDL number MFCD00043100
  • Cas Number : 106627-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Pale-yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/32289.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/32289.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart