SY3H3249F416 Synthonix Corporation

1-(5-Chloro-2-methylphenyl)propan-1-ol

  • MDL number MFCD11521240
  • Cas Number : 1314976-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37554.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37554.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart