SY3H3249F4AE Synthonix Corporation

5-Chloro-1H-indene

  • MDL number No MDL found
  • Cas Number : 3970-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/39035.html

Safety Information

MSDS https://synthonix.com/39035.html
Availability details for
Quantity
  • Availability
  • Add to Cart