SY3H3249F50A Synthonix Corporation

(4,4-Dimethyloxetan-2-yl)methanol

  • MDL number MFCD22123255
  • Cas Number : 61266-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/1980.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/1980.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart