SY3H3249F516 Synthonix Corporation

(3,3-dimethyloxetan-2-yl)methanamine

  • MDL number MFCD22666070
  • Cas Number : 34795-24-9
  • Molecular Weight : 115.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/23201.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/23201.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart