SY3H3249F52B Synthonix Corporation

(3,3-difluoropiperidin-4-yl)methanol hydrochloride

  • MDL number MFCD27978586
  • Cas Number : 1783945-29-8
  • Molecular Weight : 187.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/31244.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/31244.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart