SY3H3249F52F Synthonix Corporation

2-(2,6-Difluorophenyl)pyrrolidine

  • MDL number MFCD09809920
  • Cas Number : 1016529-46-6
  • Molecular Weight : 183.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/32071.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/32071.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart