SY3H3249F537 Synthonix Corporation

1,1-Dichloro-2-ethoxycyclopropane

  • MDL number MFCD01727254
  • Cas Number : 7363-99-7
  • Molecular Weight : 155.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/3695.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/3695.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart