SY3H3249F5AF Synthonix Corporation

2,6-Dichlorophenethyl alcohol

  • MDL number No MDL found
  • Cas Number : 30595-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/38686.html

Safety Information

MSDS https://synthonix.com/38686.html
Availability details for
Quantity
  • Availability
  • Add to Cart