SY3H3249F5CB Synthonix Corporation

5,5-Dimethylpyrrolidin-2-one

  • MDL number No MDL found
  • Cas Number : 5165-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/38848.html

Safety Information

MSDS https://synthonix.com/38848.html
Availability details for
Quantity
  • Availability
  • Add to Cart