SY3H3249F5EB Synthonix Corporation

1,6-Diazaspiro[3.4]octane-6-carboxylic acid tert-butyl ester

  • MDL number MFCD12406509
  • Cas Number : 1158749-79-1
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Viscous
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/5979.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/5979.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart