SY3H3249F5EE Synthonix Corporation

1,7-Diazaspiro[4.4]nonane-1-carboxylic acid tert-butyl ester

  • MDL number MFCD08234442
  • Cas Number : 885268-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/5985.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/5985.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart