SY3H3249F5F3 Synthonix Corporation

2,7-Diazaspiro[4.5]decane-7-carboxylic acid tert-butyl ester

  • MDL number MFCD07779071
  • Cas Number : 236406-61-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/6000.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6000.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart