SY3H3249F5F7 Synthonix Corporation

1,8-Diazaspiro[5.5]undecane-1-carboxylic acid tert-butyl ester

  • MDL number MFCD10700119
  • Cas Number : 960294-18-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/6014.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6014.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart